Learn More
Abnova™ Human HOM-TES-103 Full-length ORF (NP_542769.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_542769.2 |
---|---|
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 25900 |
Poids moléculaire | 49.3kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16107882
|
Abnova™
H00025900-P01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16197872
|
Abnova™
H00025900-P01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Alternative splicing has been observed for this gene and three transcript variants encoding different isoforms have been identified. Other alternatively spliced transcripts may exist, but their biological validity has not been confirmed. [provided by RefSeq]
Sequence: MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMRSpécification
NP_542769.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMR | |
RUO | |
IFFO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
25900 | |
HOM-TES-103 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I2223/FLJ20703/HOM-TES-103/IFFO/MGC117359 | |
IFFO1 | |
Recombinant | |
wheat germ expression system |