Learn More
Abnova™ Human HOXB8 Partial ORF (NP_076921, 16 a.a. - 114 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_076921 |
---|---|
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 3218 |
Poids moléculaire | 36.63kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16196311
|
Abnova™
H00003218-Q01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 05-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16106321
|
Abnova™
H00003218-Q01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 05-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania. [provided by RefSeq]
Sequence: GESLRPNYYDCGFAQDLGGRPTVVYGPSSGGSFQHPSQIQEFYHGPSSLSTAPYQQNPCAVACHGDPGNFYGYDPLQRQSLFGAQDPDLVQYADCKLAASpécification
NP_076921 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX2/HOX2D/Hox-2.4 | |
HOXB8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3218 | |
HOXB8 (Human) Recombinant Protein (Q01) | |
GESLRPNYYDCGFAQDLGGRPTVVYGPSSGGSFQHPSQIQEFYHGPSSLSTAPYQQNPCAVACHGDPGNFYGYDPLQRQSLFGAQDPDLVQYADCKLAA | |
RUO | |
HOXB8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |