missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSD17B12 Partial ORF (NP_057226, 203 a.a. - 271 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00051144-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16126646

  • 508.00€ / 25µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.[supplied by OMIM]

Sequence: SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNG
Spécification

Spécification

NP_057226
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNG
RUO
HSD17B12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
51144
HSD17B12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KAR/SDR12C1
HSD17B12
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions