missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human HVEM / TNFRSF14 Protein, Mouse IgG2a Fc Tag, low endotoxin

low endotoxin

Marque:  ACROBiosystems HVM-H5255-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15876947

  • 1997.41€ / 1mg
Date d’expédition estimée: 05-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Herpesvirus entry mediator (HVEM) is also known as TNFRSF14, TR2 (TNF receptorlike molecule) and ATAR (another TRAF associated receptor), is a type I membrane protein belonging to the TNF/NGF receptor superfamily. HVEM expression has been detected in peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells. The extracellular domain of HVEM has been shown to interact directly with the herpes simplex virus envelope glycoprotein D (gD). Two TNF superfamily ligands, including the secreted TNFβ (lymphotoxin α) and the membrane protein LIGHT (lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes), have been shown to be the cellular ligands for HVEM. Besides HVEM, LIGHT can also interact with LTβR, the receptor for lymphotoxin αβ heterotrimer. The role of the HVEM LIGHT /LTβ receptor ligand pair in immune function and herpesvirus pathobiology remains to be elucidated.

This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_003811.2
HEK293
Human
44.4 kDa
1 mg
Leu 39 - Val 202
TNFRSF14,ATAR,HVEA,HVEM,LIGHTR,TR2,CD270
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 0.1 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Human HVEM, Mouse IgG2a Fc Tag, low endotoxin (HVM-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 39 - Val 202 (Accession # Q92956-1).
-20°C
HVEM
Mouse IgG Fc Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions