Learn More
ACROBiosystems Human IL-11 Protein, His Tag
157.25€ - 423.38€
Spécification
Poids moléculaire | 21.0 kDa |
---|---|
Source | Human IL-11, His Tag (IL1-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20809-1). |
Immunogène | Pro 22 - Leu 199 |
Conditions de stockage | -20°C |
Alias de gène | IL-11,Interleukin-11,AGIF,Oprelvekin,IL11 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17256240
|
ACROBiosystems
IL1-H5243-20UG |
20 μg |
157.25€
20µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17246240
|
ACROBiosystems
IL1-H5243-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Interleukin-11 (IL-11) is a pleiotropic cytokine that stimulates megakaryocytopoiesis, resulting in increased production of platelets, as well as activating osteoclasts, inhibiting epithelial cell proliferation and apoptosis, and inhibiting macrophage mediator production. These functions may be particularly important in mediating the hematopoietic, osseous and mucosal protective effects of IL-11. The cytokine also possesses anti-inflammatory activity, and has been proposed as a therapeutic agent in the treatment of chronic inflammatory diseases, such as Crohn's disease and rheumatoid arthritis. Although IL-11 was initially believed to be restricted to mammals, subsequent studies demonstrated it to be expressed in fish. Despite close similarity in gene structure and conservation of key amino acids between fish and mammalian IL-11, they share relatively low overall amino acid identity and may not necessarily be functionally analogous.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.0 kDa.Spécification
21.0 kDa | |
Pro 22 - Leu 199 | |
IL-11,Interleukin-11,AGIF,Oprelvekin,IL11 | |
Unconjugated | |
His Tag | |
Powder |
Human IL-11, His Tag (IL1-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20809-1). | |
-20°C | |
IL-11 | |
Human | |
HEK293 | |
Cytokines&Growth; Factors |