Learn More
ACROBiosystems Human IL-11 Protein, His Tag
Marque:  ACROBiosystems IL1-H5243-500ug
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17266240
  • 3010.84€ / 500µg
Expédition estimée: 10-01-2023
pour voir le stock disponible 
Description
Description
Interleukin-11 (IL-11) is a pleiotropic cytokine that stimulates megakaryocytopoiesis, resulting in increased production of platelets, as well as activating osteoclasts, inhibiting epithelial cell proliferation and apoptosis, and inhibiting macrophage mediator production. These functions may be particularly important in mediating the hematopoietic, osseous and mucosal protective effects of IL-11. The cytokine also possesses anti-inflammatory activity, and has been proposed as a therapeutic agent in the treatment of chronic inflammatory diseases, such as Crohn's disease and rheumatoid arthritis. Although IL-11 was initially believed to be restricted to mammals, subsequent studies demonstrated it to be expressed in fish. Despite close similarity in gene structure and conservation of key amino acids between fish and mammalian IL-11, they share relatively low overall amino acid identity and may not necessarily be functionally analogous.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.0 kDa.
Spécification
Spécification
Powder
500 μg
Pro 22 - Leu 199
IL-11,Interleukin-11,AGIF,Oprelvekin,IL11
Unconjugated
HEK293
Human
21.0 kDa
Human IL-11, His Tag (IL1-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20809-1).
-20°C
IL-11
His Tag
Cytokines&Growth; Factors