missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-12B / NKSF2 / p40 Protein, His Tag

Marque:  ACROBiosystems NK2-H52H7-50UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877347

  • 363.17€ / 50µg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Interleukin-12 (IL12) is also known as natural killer cell stimulatory factor (NKSF), cytotoxic lymphocyte maturation factor (CLMF) , is a heterodimeric cytokine encoded by two separate genes, IL-12A (p35) and IL-12B (p40). IL12 is naturally produced by dendritic cells, macrophages and human B-lymphoblastoid cells (NC-37) in response to antigenic stimulation. IL-12 is involved in the differentiation of naive T cells into Th0 cells and plays an important role in the activities of natural killer cells and T lymphocytes.IL-12 also has anti-angiogenic activity, which means it can block the formation of new blood vessels. Interleukin-12 subunit beta (IL12B) also known as NKSF2, CLMF2 and P40. Interleukin-12 subunit beta has been shown to interact with IL23. A large excess of monomeric IL12B is also secreted by the cells producing IL12, and exhibits no demonstrable biological activity. Overexpression of IL12B gene has been shown to be associated with the pathogenesis of multiple sclerosis. In addition, studies have revealed that the promoter polymorphism of this gene is implicated in the severity of atopic and non-atopic asthma in children.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.6 kDa. The protein migrates as 44 kDa and 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH67499.1
HEK293
Human
36.6 kDa
50 µg
Ile 23 - Ser 328
IL12B,CLMF,CLMF2,NKSF,NKSF2,p40
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Cytokines and Growth Factors
Powder
>95% as determined by SDS-PAGE
Human IL-12 B, His Tag (NK2-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (Accession # P29460-1).
-20°C
IL-12 B
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions