missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-17A / CTLA-8 Protein, Tag Free

MALS verified

Marque:  ACROBiosystems ILA-H5118-25UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877067

  • 131.15€ / 25µg
Date d’expédition estimée: 05-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Interleukin-17A (IL17A) is also known as cytotoxic T-lymphocyte-associated antigen 8 (CTLA8),which is a proinflammatory cytokine produced by activated T cells. IL17A can regulate the activities of NF-kappaB and mitogen-activated protein kinases. Also,IL17A can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO).Furthermore,IL17A has been found both in glycosylated and nonglycosylated forms. High levels of IL-17 are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.

This protein carries no "e;tag"e;. The protein has a calculated MW of 15.3 kDa. the protein migrates as 18 kDa under reducing (R) condition, and 28 kDa under non-reducing (NR) condition (SDS-PAGE).
Spécification

Spécification

Less than 1.0 EU / µg by the LAL method.
Cytokines and Growth Factors
Powder
>95% as determined by SDS-PAGE
Human IL17A, Tag Free (ILA-H5118) is expressed from E.coli cells. It contains AA Gly 24 - Ala 155 (Accession # Q16552).
-20°C
IL-17 A
Tag Free
E.coli
Human
15.3 kDa
25 µg
Gly 24 - Ala 155
IL-17 A,Interleukin-17 A,CTLA-8,IL-17
Unconjugated
PBS, pH7.4
Vidéos
FDS
Documents
Certificats
Promotions

Promotions