Learn More
ACROBiosystems Human IL-1RL1 / ST2 Protein, His Tag
Marque:  ACROBiosystems IL1-H5229-200UG
Informations supplémentaires : Poids : 0.00001kg
Code produit. 15877017
  • 302.64€ / 200µg
Date d’expédition estimée: 05-01-2022
Description
Description
Interleukin 1 receptor-like 1 (IL1RL1) is also known as ST2, DER4, FIT-1, IL33R, ST2L, ST2V, which belongs to the interleukin-1 receptor family. IL1RL1 contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RL1 is receptor for interleukin-33 (IL-33), its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. IL1RL1 possibly involved in helper T-cell function. IL1RL1 can interact with MYD88, IRAK1, IRAK4, and TRAF6.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
NP_003847.2
HEK293
Human
36.9 kDa
200 µg
Lys 19 - Phe 328
IL1RL1,DER4,FIT-1,IL33R,ST2,ST2L,ST2V,T1
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>90% as determined by SDS-PAGE
Human IL-1 RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2).
-20°C
IL-1 RL1
His Tag