missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IL-1RL1 / ST2 Protein, His Tag
362.89€ - 1608.84€
Spécification
Numéro d’adhésion | NP_003847.2 |
---|---|
Poids moléculaire | 36.9 kDa |
Source | Human IL-1 RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2). |
Immunogène | Lys 19 - Phe 328 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867017
|
ACROBiosystems
IL1-H5229-1MG |
1 mg |
1608.84€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877017
|
ACROBiosystems
IL1-H5229-200UG |
200 μg |
362.89€
200µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Interleukin 1 receptor-like 1 (IL1RL1) is also known as ST2, DER4, FIT-1, IL33R, ST2L, ST2V, which belongs to the interleukin-1 receptor family. IL1RL1 contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RL1 is receptor for interleukin-33 (IL-33), its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. IL1RL1 possibly involved in helper T-cell function. IL1RL1 can interact with MYD88, IRAK1, IRAK4, and TRAF6.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_003847.2 | |
Human IL-1 RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2). | |
-20°C | |
IL-1 RL1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
36.9 kDa | |
Lys 19 - Phe 328 | |
IL1RL1,DER4,FIT-1,IL33R,ST2,ST2L,ST2V,T1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |