missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-36 beta (IL-1F8) Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 200-36B-2UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17890073

  • 78.00€ / 2µg
Expédition estimée: 13-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.
Spécification

Spécification

Q9NZH7
protein with no preservative
17.3
2 µg
-20°C
<1 EU/µg
IL36B
Unconjugated
MREAAPKSYA IRDSRQMVWV LSGNSLIAAP LSRSIKPVTL HLIACRDTEF SDKEKGNMVY LGIKGKDLCL FCAEIQGKPT LQLKEKNIMD LYVEKKAQKP FLFFHNKEGS TSVFQSVSYP GWFIATSTTS GQPIFLTKER GITNNTNFYL DSVE
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
27177
Human IL-36 beta (IL-1F8)
E. coli
RUO
2310043N20Rik; family of interleukin 1-eta; FIL1; FIL1 eta; FIL1-(ETA); Fil1e; FIL1H; FILI-(ETA); IL-1 eta; IL1-ETA; IL1F8; IL-1F8; IL1F8 (Canonical product IL-1F8a); IL-1F8 (FIL1-eta); IL1H2; IL-1H2; Il36b; interleukin 1 family, member 8; interleukin 1 family, member 8 (eta); interleukin 1, eta; interleukin 36, beta; interleukin-1 eta; Interleukin-1 family member 8; interleukin-1 homolog 2; Interleukin-1 Superfamily e; interleukin-36 beta
Human
Recombinant
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions