missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein, His Tag&Tag Free
495.95€ - 3870.90€
Spécification
Numéro d’adhésion | NP 003629.2 (ITGA8) and NP_002202.2 (ITGB1) |
---|---|
Poids moléculaire | 114.6 kDa (ITGA8) and 83.7 kDa (ITGB1) |
Source | Human ITGA8&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) and P05556-1 (ITGB1)). |
Immunogène | Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857147
|
ACROBiosystems
IT1-H52W9-100UG |
100 μg |
495.95€
100µg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867147
|
ACROBiosystems
IT1-H52W9-1MG |
1 mg |
3870.90€
1mg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Human Integrin alpha 8 beta 1 Heterodimer Protein consists of ITGA8 and ITGB1. The integrin alpha 8 subunit, isolated by low stringency hybridization, is a novel integrin subunit that associates with beta 1. The recently identified alpha 8 integrin subunit associates with beta 1 and is predominantly expressed in smooth muscle and other contractile cells in adult tissues, and in mesenchymal and neural cells during development. In addition, Integrin alpha 8 beta 1 is a receptor for fibronectin and can promote attachment, cell spreading, and neurite outgrowth on fibronectin.
Human ITGA8&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 114.6 kDa (ITGA8) and 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistidSpécification
NP 003629.2 (ITGA8) and NP_002202.2 (ITGB1) | |
Human ITGA8&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) and P05556-1 (ITGB1)). | |
-20°C | |
Integrin alpha 8 beta 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
114.6 kDa (ITGA8) and 83.7 kDa (ITGB1) | |
Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) | |
Integrin alpha 8 beta 1,ITGA8andITGB1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |