Learn More
Abnova™ Human INTS4 Full-length ORF (NP_291025.3, 1 a.a. - 963 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00092105-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM]
Sequence: MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARRSpécification
NP_291025.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
134.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
INT4/MGC16733/MST093 | |
INTS4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
92105 | |
INTS4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARR | |
RUO | |
INTS4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |