missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IQCB1 Full-length ORF (NP_001018865.1, 1 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

sur devis

Spécification

Nom IQCB1 (Human) Recombinant Protein (P01)
Recombinant Yes
Méthode de purification Glutathione Sepharose 4 Fast Flow
Poids moléculaire 80.2kDa
Conditions de stockage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Consulter d'autres spécifications

Description

Description

Sequence: MKPTGTDPRILSIAAEVAKSPEQNVPVILLKLKEIINITPLGSSELKKIKQDIYCYDLIQYCLLVLSQDYSRIQGGWTTISQLTQILSHCCVGLEPGEDAEEFYNELLPSAAENFLVLGRQLQTCFINAAKAEEKDELLHFFQIVTDSLFWLLGGHVELIQNVLQSDHFLHLLQADNVQIGSAVMMMLQNILQINRSKRSKMLLEINRQKEEEDLKLQLQLQRQRAMRLSRELQLSMLEIVHPGQVEKHYREMEEKSALIIQKHWRGYRERKNFHQQRQSLIEYKAAVTLQRAALKFLAKYRKKKKLFAPWRGLQELTDARRVELKKRVDDYVRRHLGSPMSDVVSRELHAQAQERLQHYFMGRALEERAQQHREALIAQISTNVEQLMKAPSLKEAEGKEPELFLSRSRPVAAKAKQAHLTTLKHIQAPWWKKLGEESGDEIDVPKDELSIELENLFIGGTKPP
Spécification

Spécification

IQCB1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
80.2kDa
Vidéos
FDS
Documents

Documents

Product Certifications