missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KIAA1604 Full-length ORF (AAH93952.1, 1 a.a. - 908 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
sur devis
Spécification
Nom | KIAA1604 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Méthode de purification | Glutathione Sepharose 4 Fast Flow |
Poids moléculaire | 131.9kDa |
Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
Sequence: MKSSVAQIKPSSGHDRRENLNSYQRNSSPEDRYEEQERSPRDRDYFDYSRSDYEHSRRGRSYDSSMESRNRDREKRRERERDTDRKRSRKSPSPGRRNPETSVTQSSSAQDEPATKKKKDELDPLLTRTGGAYIPPAKLRMMQEQITDKNSLAYQRMSWEALKKSINGLINKVNISNISIIIQELLQENIVRGRGLLSRSVLQAQSASPIFTHVYAALVAIINSKFPQIGELILKRLILNFRKGYRRNDKQLCLTASKFVAHLINQNVAHEVLCLEMLTLLLERPTDDSVEVAIGFLKECGLKLTQVSPRGINAIFERLRNILHESEIDKRVQYMIEVMFAVRKDGFKDHPIILEGLDLVEEDDQFTHMLPLEDDYNPEDVLNVFKMDPNFMENEEKYKAIKKEILDEGDTDSNTDQDAGSSEEDEEEEEEEGEEDEEGQKVTIHDKTEINLVSFRRTIYLAIQSSLDFEECAHKLLKMEFPESQTKELCNMILDCCAQQRTYEKFFGLLAGRFCMLKKEYMESFEGIFKEQYDTIHRLETNKLRNVAKMFAHLLYTDSLPWSVLECIKLSEETTTSSSRIFVKIFFQELCEYMGLPKLNARLKDETLQPFFEGLLPRDNPRNTRFAINFFTSIGLGGLTDELREHLKNTPKVIVAQKPDVEQNKSSPSSSSSASSSSESDSSDSDSDSSDSSSESSSEESDSSSISSHSSASANDVRKKGHGKTRSKEVDKLIRNQQTNDRKQKERRQEHGHQETRTERERRSEKHRDQNSRGSNWRDPITKYTSDKDVPSERNNYSRVANDRDQEMHIDLENKHGDPKKKRGERRNSFSENEKHTHRIKDSENFRRKDRSKSKEMNRKHSGSRSDEDRYQNGAERRWEKSSRYSEQSRESKKNQDRRREKSPAKQKSpécification
KIAA1604 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
131.9kDa |