missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1604 Full-length ORF (AAH93952.1, 1 a.a. - 908 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00057703-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122923

  • 295.00€ / 10µg

Expédition estimée
30-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MKSSVAQIKPSSGHDRRENLNSYQRNSSPEDRYEEQERSPRDRDYFDYSRSDYEHSRRGRSYDSSMESRNRDREKRRERERDTDRKRSRKSPSPGRRNPETSVTQSSSAQDEPATKKKKDELDPLLTRTGGAYIPPAKLRMMQEQITDKNSLAYQRMSWEALKKSINGLINKVNISNISIIIQELLQENIVRGRGLLSRSVLQAQSASPIFTHVYAALVAIINSKFPQIGELILKRLILNFRKGYRRNDKQLCLTASKFVAHLINQNVAHEVLCLEMLTLLLERPTDDSVEVAIGFLKECGLKLTQVSPRGINAIFERLRNILHESEIDKRVQYMIEVMFAVRKDGFKDHPIILEGLDLVEEDDQFTHMLPLEDDYNPEDVLNVFKMDPNFMENEEKYKAIKKEILDEGDTDSNTDQDAGSSEEDEEEEEEEGEEDEEGQKVTIHDKTEINLVSFRRTIYLAIQSSLDFEECAHKLLKMEFPESQTKELCNMILDCCAQQRTYEKFFGLLAGRFCMLKKEYMESFEGIFKEQYDTIHRLETNKLRNVAKMFAHLLYTDSLPWSVLECIKLSEETTTSSSRIFVKIFFQELCEYMGLPKLNARLKDETLQPFFEGLLPRDNPRNTRFAINFFTSIGLGGLTDELREHLKNTPKVIVAQKPDVEQNKSSPSSSSSASSSSESDSSDSDSDSSDSSSESSSEESDSSSISSHSSASANDVRKKGHGKTRSKEVDKLIRNQQTNDRKQKERRQEHGHQETRTERERRSEKHRDQNSRGSNWRDPITKYTSDKDVPSERNNYSRVANDRDQEMHIDLENKHGDPKKKRGERRNSFSENEKHTHRIKDSENFRRKDRSKSKEMNRKHSGSRSDEDRYQNGAERRWEKSSRYSEQSRESKKNQDRRREKSPAKQK
Spécification

Spécification

AAH93952.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
131.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EIF4GL|KIAA1604|NCM|fSAPb
CWC22
Yes
wheat germ expression system
Liquid
57703
KIAA1604 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKSSVAQIKPSSGHDRRENLNSYQRNSSPEDRYEEQERSPRDRDYFDYSRSDYEHSRRGRSYDSSMESRNRDREKRRERERDTDRKRSRKSPSPGRRNPETSVTQSSSAQDEPATKKKKDELDPLLTRTGGAYIPPAKLRMMQEQITDKNSLAYQRMSWEALKKSINGLINKVNISNISIIIQELLQENIVRGRGLLSRSVLQAQSASPIFTHVYAALVAIINSKFPQIGELILKRLILNFRKGYRRNDKQLCLTASKFVAHLINQNVAHEVLCLEMLTLLLERPTDDSVEVAIGFLKECGLKLTQVSPRGINAIFERLRNILHESEIDKRVQYMIEVMFAVRKDGFKDHPIILEGLDLVEEDDQFTHMLPLEDDYNPEDVLNVFKMDPNFMENEEKYKAIKKEILDEGDTDSNTDQDAGSSEEDEEEEEEEGEEDEEGQKVTIHDKTEINLVSFRRTIYLAIQSSLDFEECAHKLLKMEFPESQTKELCNMILDCCAQQRTYEKFFGLLAGRFCMLKKEYMESFEGIFKEQYDTIHRLETNKLRNVAKMFAHLLYTDSLPWSVLECIKLSEETTTSSSRIFVKIFFQELCEYMGLPKLNARLKDETLQPFFEGLLPRDNPRNTRFAINFFTSIGLGGLTDELREHLKNTPKVIVAQKPDVEQNKSSPSSSSSASSSSESDSSDSDSDSSDSSSESSSEESDSSSISSHSSASANDVRKKGHGKTRSKEVDKLIRNQQTNDRKQKERRQEHGHQETRTERERRSEKHRDQNSRGSNWRDPITKYTSDKDVPSERNNYSRVANDRDQEMHIDLENKHGDPKKKRGERRNSFSENEKHTHRIKDSENFRRKDRSKSKEMNRKHSGSRSDEDRYQNGAERRWEKSSRYSEQSRESKKNQDRRREKSPAKQK
RUO
CWC22
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents