missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human LAIR1 / CD305 Protein, His Tag

419.88€ - 2927.16€

Spécification

Numéro d’adhésion AAH27899.1
Poids moléculaire 17.4 kDa
Source Human LAIR-1, His Tag (CD5-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GT x 8-1).
Immunogène Gln 22 - His 163
Conditions de stockage -20°C
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15876207
Afficher les documents Détails de la promotion
ACROBiosystems
CD5-H52H1-100UG
100 μg
419.88€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15886207
Afficher les documents Détails de la promotion
ACROBiosystems
CD5-H52H1-1MG
1 mg
2927.16€
1mg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Leukocyte-associated immunoglobulin-like receptor-1 (LAIR-1) is constitutively expressed on the majority of human peripheral blood mononuclear leukocytes. LAIR-1 or CD305 is a transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs. LAIR-1 recruits SHP-1 and SHP-2 phosphatases upon activation, and cross-linking of the LAIR-1 antigen on natural killer (NK) cells results in strong inhibition of NK cell–mediated cytotoxicity. Functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation.Diseases associated with LAIR1 include Chronic Active Epstein-Barr Virus Infection and Palindromic Rheumatism.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.4 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH27899.1
Human LAIR-1, His Tag (CD5-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GT x 8-1).
-20°C
LAIR-1
Human
PBS, pH7.4
HEK293
Immune Checkpoint Proteins
17.4 kDa
Gln 22 - His 163
LAIR1,CD305
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by reduced SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications