missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human LILRB1 / CD85j / ILT2 Protein, Fc Tag

299.92€ - 2135.39€

Spécification

Numéro d’adhésion AAH15731.1
Poids moléculaire 73.8 kDa
Source Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM8-1).
Immunogène Gly 24 - His 458
Conditions de stockage -20°C
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15866407
Afficher les documents Détails de la promotion
ACROBiosystems
CDJ-H5252-1MG
1 mg
2135.39€
1mg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15856407
Afficher les documents Détails de la promotion
ACROBiosystems
CDJ-H5252-100UG
100 μg
299.92€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

CD85 antigen-like family member J (CD85J) is also known as Leukocyte immunoglobulin-like receptor subfamily B member 1 (LILRB1 or LIR-1), Immunoglobulin-like transcript 2 (ILT-2), Monocyte/macrophage immunoglobulin-like receptor 7 (MIR7), which belongs to leukocyte immunoglobulin-like receptor (LIR) family. CD85J / LILRB1 Contains 4 Ig-like C2-type (immunoglobulin-like) domains. CD85J / LIR-1 is expressed predominantly on B-cells and monocytes. CD85J is receptor for class I MHC antigens and recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles and is also receptor for H301/UL18, a human cytomegalovirus class I MHC homolog. Ligand binding results in inhibitory signals and down-regulation of the immune response. CD85J / LILRB1 interaction with HLA-B or HLA-E leads to inhibition of the signal triggered by FCER1A and inhibits serotonin release. CD85J / LILRB1 inhibits FCGR1A-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH15731.1
Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM8-1).
-20°C
LILRB1
Human
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Immune Checkpoint Proteins
73.8 kDa
Gly 24 - His 458
CD85J,LILRB1,CD85,ILT2,LIR1,MIR7
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>92% as determined by SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications