Learn More
ACROBiosystems Human LILRB5 / CD85c / LIR-8 Protein, His Tag
Marque: ACROBiosystems CDC-H5220-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Leukocyte immunoglobulin-like receptor subfamily B member 5 (LILRB5), also known as CD85c and LIR-8, belongs to the leukocyte immunoglobulin-like receptor (LIR) family of transmembrane glycoproteins that negatively regulate immune cell activation. Several other LIR subfamily B receptors are expressed on immune cells where they bind to MHC class I molecules on antigen-presenting cells and inhibit stimulation of an immune response.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 66-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_006831 | |
100 μg | |
Gly 24 - Gly 458 | |
LILRB5,CD85c,LIR-8 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
49.2 kDa | |
Human LILRB5, His Tag (CDC-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # NP_006831). | |
-20°C | |
LILRB5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |