Learn More
Abnova™ Human LMCD1 Partial ORF (NP_055398, 266 a.a. - 364 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00029995-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene contains a cysteine-rich domain in the N-terminal region and 2 LIM domains in the C-terminal region. It also has several potential phosphorylation and N-myristoylation sites and a single potential N-glycosylation site. The presence of LIM domains implies involvement in protein-protein interactions. Expression of this gene has been detected in most tissues, with highest expression in skeletal muscle. Transcript variants utilizing alternative polyA signals have been observed. [provided by RefSeq]
Sequence: KQWHPTCFVCAKCSEPLVDLIYFWKDGAPWCGRHYCESLRPRCSGCDEIIFAEDYQRVEDLAWHRKHFVCEGCEQLLSGRAYIVTKGQLLCPTCSKSKRSpécification
NP_055398 | |
Liquid | |
29995 | |
LMCD1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMCD1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KQWHPTCFVCAKCSEPLVDLIYFWKDGAPWCGRHYCESLRPRCSGCDEIIFAEDYQRVEDLAWHRKHFVCEGCEQLLSGRAYIVTKGQLLCPTCSKSKR | |
RUO | |
LMCD1 | |
Yes | |
wheat germ expression system |