missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MACF1 Partial ORF (AAH07330, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00023499-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16127382

  • 508.00€ / 25µg
Expédition estimée: 07-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene belongs to the plakin family of cytoskeletal linker proteins. This protein family forms bridges between different cytoskeletal elements through specialized modular domains. The encoded protein is one of the largest size proteins identified in human cytoskeletal proteins. It has functional actin and microtubule binding domains, and it appears to stabilize actin at sites where microtubules and microfilaments meet. It may function in microtubule dynamics to facilitate actin-microtubule interactions at the cell periphery and to couple the microtubule network to cellular junctions. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: KIEDEVTRQVAQCKCAKRFQVEQIGENKYRFGDSQQLRLVRILRSTVMVRVGGGWMALDEFLVKNDPCRARGRTNIELREKFILPEGASQGMTPF
Spécification

Spécification

AAH07330
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KIEDEVTRQVAQCKCAKRFQVEQIGENKYRFGDSQQLRLVRILRSTVMVRVGGGWMALDEFLVKNDPCRARGRTNIELREKFILPEGASQGMTPF
RUO
MACF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23499
MACF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABP620/ACF7/FLJ45612/FLJ46776/KIAA0465/KIAA1251/MACF/OFC4
MACF1
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions