missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human MMP-2 Protein, His Tag

Marque:  ACROBiosystems MM2-H5225-50UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877307

  • 252.20€ / 50µg
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Matrix metalloproteinase-2 (MMP-2) is also known as 72 kDa type IV collagenase, 72 kDa gelatinase, Gelatinase A and CLG4A, which belongs to the peptidase M10A family. MMP-2 / CLG4A contains 3 fibronectin type-II domains and 4 hemopexin-like domains. MMP-2 is produced by normal skin fibroblasts. MMP-2 cleaves the collagen-like sequence Pro-Gln-Gly-|-Ile-Ala-Gly-Gln. MMP2 involved in diverse functions such as remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, and atherosclerotic plaque rupture. As well as degrading extracellular matrix proteins, can also act on several nonmatrix proteins such as big endothelial 1 and beta-type CGRP promoting vasoconstriction. Also cleaves KISS at a Gly-|-Leu bond. Appears to have a role in myocardial cell death pathways. Contributes to myocardial oxidative stress by regulating the activity of GSK3beta. Cleaves GSK3beta in vitro. PEX, the C-terminal non-catalytic fragment of MMP2, posseses anti-angiogenic and anti-tumor properties and inhibits cell migration and cell adhesion to FGF2 and vitronectin.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.8 kDa. The protein migrates as 66-71 kDa under reducing (R) condition (SDS-PAGE).
Spécification

Spécification

AAH02576
HEK293
Human
71.8 kDa
50 µg
Ala 30 - Cys 660
MMP2,CLG4,CLG4A,MMP-II,MONA,TBE-1
Unconjugated
50 mM Tris, 150 mM NaCl, pH8.0
Less than 1.0 EU / µg by the LAL method.
Enzymes
Powder
>92% as determined by SDS-PAGE
Human MMP-2, His Tag (MM2-H5225) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Cys 660 (Accession # AAH02576).
-20°C
MMP-2
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions