missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MVD Partial ORF (NP_002452, 301 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00004597-Q01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16121495

  • 310.00€ / 10µg

Expédition estimée
11-01-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: AYTFDAGPNAVIFTLDDTVAEFVAAVWHGFPPGSNGDTFLKGLQVRPAPLSAELQAALAMEPTPGGVKYIIVTQVGPGPQILDDPCAHLLGPDGLPKP
Spécification

Spécification

NP_002452
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AYTFDAGPNAVIFTLDDTVAEFVAAVWHGFPPGSNGDTFLKGLQVRPAPLSAELQAALAMEPTPGGVKYIIVTQVGPGPQILDDPCAHLLGPDGLPKP
RUO
MVD
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4597
MVD (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FP17780|MPD
MVD
Yes
Vidéos
FDS
Documents