missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human NKp30 / NCR3 / CD337 Protein, His Tag

Marque:  ACROBiosystems NC3-H5228-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877337

  • 1815.83€ / 1mg
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Natural cytotoxicity triggering receptor 3 (NCR3) is also known as Activating natural killer receptor p30, Natural killer cell p30-related protein (NK-p30), CD antigen CD337, 1C7, LY117. NCR3 /CD337 /NKp30 belongs to the natural cytotoxicity receptor (NCR) family. NCR3 /NKp30 contains one Ig-like (immunoglobulin-like) domain. NCR3 /NKp30 is selectively expressed by all resting and activated NK cells and weakly expressed in spleen. NCR3 is homodimer in the unliganted form. NCR3 interacts with CD3Z. NCR3 interacts with and is activated by binding to NCR3LG1 or BAG6. Engagement of NCR3 by BAG6 also promotes dendritic cell (DC) maturation, both through killing those DCs that did not properly acquire a mature phenotype, and inducing NK cells to release TNFA and IFNG, which promotes DC maturation.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.7 kDa. The protein migrates as 21-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH52582
HEK293
Human
13.7 kDa
1 mg
Leu 19 - Gly 135
NCR3,CD337,NKp30,1C7,LY117,MALS
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582).
-20°C
NKp30
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions