Learn More
ACROBiosystems Human NKp30 / NCR3 / CD337 Protein, His Tag
374.99€ - 2419.31€
Spécification
Numéro d’adhésion | AAH52582 |
---|---|
Poids moléculaire | 13.7 kDa |
Source | Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582). |
Immunogène | Leu 19 - Gly 135 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867337
|
ACROBiosystems
NC3-H5228-100UG |
100 μg |
374.99€
100µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877337
|
ACROBiosystems
NC3-H5228-1MG |
1 mg |
2419.31€
1mg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Natural cytotoxicity triggering receptor 3 (NCR3) is also known as Activating natural killer receptor p30, Natural killer cell p30-related protein (NK-p30), CD antigen CD337, 1C7, LY117. NCR3 /CD337 /NKp30 belongs to the natural cytotoxicity receptor (NCR) family. NCR3 /NKp30 contains one Ig-like (immunoglobulin-like) domain. NCR3 /NKp30 is selectively expressed by all resting and activated NK cells and weakly expressed in spleen. NCR3 is homodimer in the unliganted form. NCR3 interacts with CD3Z. NCR3 interacts with and is activated by binding to NCR3LG1 or BAG6. Engagement of NCR3 by BAG6 also promotes dendritic cell (DC) maturation, both through killing those DCs that did not properly acquire a mature phenotype, and inducing NK cells to release TNFA and IFNG, which promotes DC maturation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.7 kDa. The protein migrates as 21-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH52582 | |
Human NKp30, His Tag (NC3-H5228) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Gly 135 (Accession # AAH52582). | |
-20°C | |
NKp30 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
13.7 kDa | |
Leu 19 - Gly 135 | |
NCR3,CD337,NKp30,1C7,LY117,MALS | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |