Learn More
ACROBiosystems Human NKp46 / NCR1 / CD335 Protein, Fc Tag
MALS verified
Marque: ACROBiosystems NC1-H5257-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Natural cytotoxicity triggering receptor 1 (NCR1) is also known as Natural killer cell p46-related protein (NK-p46), Lymphocyte antigen 94 homolog (LY94), CD antigen CD335, which belongs to the natural cytotoxicity receptor (NCR) family. NCR1 contains two Ig-like (immunoglobulin-like) domains. NCR1 interacts with CD247 and FCER1G. NCR1 / CD335 may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.0 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH64806 | |
1 mg | |
Gln 22 - Asn 254 | |
NCR1,LY94,CD335,NK-p46,hNKp46 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
53.0 kDa | |
Human NKp46, Fc Tag (NC1-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806). | |
-20°C | |
NKp46 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |