Learn More
Abnova™ Human NPR2 Partial ORF (NP_003986.2, 131 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00004882-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide (CNP), which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type. [provided by RefSeq]
Sequence: AKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPLSpécification
NP_003986.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPL | |
RUO | |
NPR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4882 | |
NPR2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMDM/ANPRB/GUC2B/GUCY2B/NPRB/NPRBi | |
NPR2 | |
Recombinant | |
wheat germ expression system |