Learn More
Abnova™ Human OTUB2 Full-length ORF (NP_075601.1, 1 a.a. - 234 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00078990-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
Otubains are deubiquitylating cysteine proteases (DUBs; see MIM 602519) that belong to the ovarian tumor (OTU) protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin (UB; see MIM 191339)-protein bond.[supplied by OMIM]
Sequence: MSETSFNLISEKCDILSILRDHPENRIYRRKIEELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNFFNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDFCTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNILYAADKHSpécification
NP_075601.1 | |
Liquid | |
78990 | |
OTUB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSETSFNLISEKCDILSILRDHPENRIYRRKIEELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNFFNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDFCTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNILYAADKH | |
RUO | |
OTUB2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C14orf137/FLJ21916/MGC3102/OTB2/OTU2 | |
OTUB2 | |
Yes | |
wheat germ expression system |