Learn More
ACROBiosystems Human P-Selectin / CD62P Protein, Fc Tag
314.52€ - 1959.64€
Spécification
Numéro d’adhésion | AAN06828 |
---|---|
Poids moléculaire | 106.6 kDa |
Source | Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (HEK293). It contains AA Trp 42 - Ala 771 (Accession # AAN06828). |
Immunogène | Trp 42 - Ala 771 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15877677
|
ACROBiosystems
SEP-H5255-1MG |
1 mg |
1959.64€
1mg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867677
|
ACROBiosystems
SEP-H5255-100UG |
100 μg |
314.52€
100µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
P-selectin (SELP) is also known as CD62 antigen-like family member P, granule membrane protein 140 (GMP-140), leukocyte-endothelial cell adhesion molecule 3 (LECAM3) and platelet activation dependent granule-external membrane protein (PADGEM). SELP functions as a cell adhesion molecule (CAM) on the surfaces of activated endothelial cells, which line the inner surface of blood vessels, and activated platelets. In unactivated endothelial cells, it is stored in granules called Weibel-Palade bodies. In unactivated platelets SELP is stored in α-granules. The primary ligand for SELP is P-selectin glycoprotein ligand-1 (PSGL-1) which is expressed on almost all leukocytes, although P-selectin also binds to heparan sulfate and fucoidan. Furthermore, SELP has a functional role in metastasis of tumor similar to E-selectin, which can help cancer cells invade into bloodstream for metastasis and provided locally with multiple growth factors respectively.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 106.6 kDa. The protein migrates as 146-160 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Spécification
AAN06828 | |
Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (HEK293). It contains AA Trp 42 - Ala 771 (Accession # AAN06828). | |
-20°C | |
P-Selectin | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
106.6 kDa | |
Trp 42 - Ala 771 | |
P-Selectin,CD62P,SELP,GMP-140 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |