missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRICKLE1 Partial ORF (NP_694571, 684 a.a. - 756 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00144165-Q01.10ug

Additional Details : Poids : 0.00010kg

 View more versions of this product

Product Code. 16121997

  • 335.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a nuclear receptor that may be a negative regulator of the Wnt/beta-catenin signaling pathway. The encoded protein localizes to the nuclear membrane and has been implicated in the nuclear trafficking of the transcription repressors REST/NRSF and REST4. Mutations in this gene have been linked to progressive myoclonus epilepsy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3. [provided by RefSeq]

Sequence: DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG
Specifications

Specifications

NP_694571
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG
RUO
PRICKLE1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
144165
PRICKLE1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPM1B/FLJ31627/FLJ31937/MGC138902/MGC138903/RILP
PRICKLE1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers