Learn More
Abnova™ Human PRPH Partial ORF (NP_006253, 374 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00005630-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a cytoskeletal protein found in neurons of the peripheral nervous system. The encoded protein is a type III intermediate filament protein with homology to other cytoskeletal proteins such as desmin, and is a different protein that the peripherin found in photoreceptors. Mutations in this gene have been associated with susceptibility to amyotrophic lateral sclerosis. [provided by RefSeq]
Sequence: RHLREYQELLNVKMALDIEIATYRKLLEGEESRISVPVHSFASLNIKTTVPEVEPPQDSHSRKTVLIKTIETRNGEVVTESQKEQRSELDKSSAHSYSpécification
NP_006253 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RHLREYQELLNVKMALDIEIATYRKLLEGEESRISVPVHSFASLNIKTTVPEVEPPQDSHSRKTVLIKTIETRNGEVVTESQKEQRSELDKSSAHSY | |
RUO | |
PRPH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5630 | |
PRPH (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NEF4/PRPH1 | |
PRPH | |
Recombinant | |
wheat germ expression system |