Learn More
Abnova™ Human RAI2 Full-length ORF (NP_068557.1, 1 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010742-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function of this retinoic acid-induced gene has not yet been determined; however, it has been suggested to play a role in development. Localization of this gene designates it to be a candidate for diseases such as Nance-Horan syndrome, sensorineural deafness, non-specific X-linked mental retardation, oral-facial-digital syndrome, and Fried syndrome. [provided by RefSeq]
Sequence: MDDLQSQNLSMDMTDSPPALANNRLENGMAQLITTEAWNINSTDLVKKALVTVPAPSILNPPAESQSGMALKVAATVLQPLCLGESPVVMPIHMQVEGSSAPELNPNGNATYVMTTQGPVQLPVVLEQHVFQHLNSPLVLPQEAPCSSSTIHNNLFQGAEDPEAQPQLLDLRIPSQPQEPTLPFEAVLQNLFPSQGTLGPPPCQPPPGYAPVPPQPFSSPLSPLVPPATLLVPYPVIVPLPVPVPIPIPIPVPQSSESKFSSSFPKPPSSFGLHPFKGTQTPLEKDELKPFDILQPKEYFQLSRHTVIKMGSENEALDLSMKSVPWLKAGEVSPPIFQEDAALDLSVAAHRKSEPPPETLYDSGASVDSSGHTVMEKLPSGMEISFAPATSHEAPAMMDSHISSSDAATEMLSQPNHPSGEVKAENNIEMVGESQAAKVIVSVEDAVPTIFCGKIKGLSGVSTKNFSFKREDSVLQGYDINSQGEESMGNAEPLRKPIKNRSIKLKKVNSQEIHMLPIKKQRLATFFPRKSpécification
NP_068557.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10742 | |
RAI2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDDLQSQNLSMDMTDSPPALANNRLENGMAQLITTEAWNINSTDLVKKALVTVPAPSILNPPAESQSGMALKVAATVLQPLCLGESPVVMPIHMQVEGSSAPELNPNGNATYVMTTQGPVQLPVVLEQHVFQHLNSPLVLPQEAPCSSSTIHNNLFQGAEDPEAQPQLLDLRIPSQPQEPTLPFEAVLQNLFPSQGTLGPPPCQPPPGYAPVPPQPFSSPLSPLVPPATLLVPYPVIVPLPVPVPIPIPIPVPQSSESKFSSSFPKPPSSFGLHPFKGTQTPLEKDELKPFDILQPKEYFQLSRHTVIKMGSENEALDLSMKSVPWLKAGEVSPPIFQEDAALDLSVAAHRKSEPPPETLYDSGASVDSSGHTVMEKLPSGMEISFAPATSHEAPAMMDSHISSSDAATEMLSQPNHPSGEVKAENNIEMVGESQAAKVIVSVEDAVPTIFCGKIKGLSGVSTKNFSFKREDSVLQGYDINSQGEESMGNAEPLRKPIKNRSIKLKKVNSQEIHMLPIKKQRLATFFPRK | |
RUO | |
RAI2 | |
Recombinant | |
wheat germ expression system |