Learn More
Abnova™ Human RANBP3 Full-length ORF (AAH04349.1, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00008498-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein with a RanBD1 domain that is found in both the nucleus and cytoplasm. This protein plays a role in nuclear export as part of a heteromeric complex. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MADLANEEKPAIAPPVFVFQKDKGQKRSAGGSSPEGGEDSDREDGNYCPPVKRERTSSLTQFPPSQSEERSSGFRLKPPTLIHGQAPSAGLPSQKPKEQQRSVLRPAVLQAPQPKALSQTVPSSGTNGVSLPADCTGAVPAASPDTAAWRSPSEAADEVCALEEKEPQKNESSNASEEEACEKKDPATQQAFVFGQNLRDRVKLINESVDEADMENAGHPSADTPTATNYFLQYISSSLENSTNSADASSNKFVFGQNMSERVLSPPKLNEVSSDANRENAAAESGSESSSQEATPEKESLAESAAAYTKATARKCLLEKVEVITGEEAESNVLQMQCKLFVFDKTSQSWLLRHPHPSHPAVGPCLCGEQPALCPCPGLPNYRPGTPAQLGGLLVRVSpécification
AAH04349.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I1520 | |
RANBP3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8498 | |
RANBP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADLANEEKPAIAPPVFVFQKDKGQKRSAGGSSPEGGEDSDREDGNYCPPVKRERTSSLTQFPPSQSEERSSGFRLKPPTLIHGQAPSAGLPSQKPKEQQRSVLRPAVLQAPQPKALSQTVPSSGTNGVSLPADCTGAVPAASPDTAAWRSPSEAADEVCALEEKEPQKNESSNASEEEACEKKDPATQQAFVFGQNLRDRVKLINESVDEADMENAGHPSADTPTATNYFLQYISSSLENSTNSADASSNKFVFGQNMSERVLSPPKLNEVSSDANRENAAAESGSESSSQEATPEKESLAESAAAYTKATARKCLLEKVEVITGEEAESNVLQMQCKLFVFDKTSQSWLLRHPHPSHPAVGPCLCGEQPALCPCPGLPNYRPGTPAQLGGLLVRV | |
RUO | |
RANBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |