Learn More
ACROBiosystems Human RANK / TNFRSF11A Protein, Mouse IgG2a Fc Tag, low endotoxin
MALS verified
423.38€ - 2818.49€
Spécification
Numéro d’adhésion | NP_003830.1 |
---|---|
Poids moléculaire | 47.2 kDa |
Source | Human RANK, Mouse IgG2a Fc Tag, low endotoxin (RAK-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Pro 212 (Accession # Q9Y6Q6-1). |
Immunogène | Ile 30 - Pro 212 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867577
|
ACROBiosystems
RAK-H5251-1MG |
1 mg |
2818.49€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15857577
|
ACROBiosystems
RAK-H5251-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
RANK / TNFRSF11A is a member of the tumor necrosis factor receptor family. RANK / TNFRSF11A shares significant amino acid homology with other members of the TNF R family in its extracellular four cysteine-rich repeats. Functional genomics in mice showed that RANKL (TNFSF11), a member of the TNF ligand superfamily, and RANK (TNFRSF11a), the cognate TNFR family receptor for RANKL, were essential for osteoclastogenesis in vivo. A functional interaction between RANKL, expressed by bone stromal cells of the osteoblast lineage, and RANK, expressed by osteoclast precursors of hematopoietic myeloid lineage, is necessary for osteoclast differentiation, survival, and activation.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 47.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_003830.1 | |
Human RANK, Mouse IgG2a Fc Tag, low endotoxin (RAK-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Pro 212 (Accession # Q9Y6Q6-1). | |
-20°C | |
RANK | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
47.2 kDa | |
Ile 30 - Pro 212 | |
CD265,FEO,LOH18CR1,ODFR,OFE,OPTB7,OSTS,PDB2,RANK,TNFRSF11A,TRANCER,CD265 antigen,ODFROSTS,RANKLOH18CR1,NFKB activator | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |