Learn More
Abnova™ Human RNF40 Partial ORF (NP_055586, 102 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00009810-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery. [provided by RefSeq]
Sequence: DETVEALLRCHESQGELSSAPEAPGTQEGPTCDGTPLPEPGTSELRDPLLMQLRPPLSEPALAFVVALGASSSEEVELELQGRMEFSKAAVSRVVEASDSpécification
NP_055586 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DETVEALLRCHESQGELSSAPEAPGTQEGPTCDGTPLPEPGTSELRDPLLMQLRPPLSEPALAFVVALGASSSEEVELELQGRMEFSKAAVSRVVEASD | |
RUO | |
RNF40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9810 | |
RNF40 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRE1B/DKFZp686K191/KIAA0661/MGC13051/RBP95/STARING | |
RNF40 | |
Recombinant | |
wheat germ expression system |