Learn More
ACROBiosystems Human ROR2 / NTRKR2 Protein, His Tag
Marque:  ACROBiosystems RO2-H52E5-50UG
Informations supplémentaires : Poids : 0.00001kg
 Afficher plus de versions de ce produit
Code produit. 15867607
  • 443.87€ / 50µg
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Description
Description
Tyrosine-protein kinase transmembrane receptor ROR2 is also known as Neurotrophic tyrosine kinase, receptor-related 2 (NTRKR2), which belongs to the protein kinase superfamily and Tyr protein kinase family and ROR subfamily. ROR2 is a homodimer protein, which can binds YWHAB or interact with WTIP. ROR2 may be involved in the early formation of the chondrocytes. It seems to be required for cartilage and growth plate development.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
NP_004551.1
50 μg
Glu 34 - Gly 403
ROR2,NTRKR2
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
43.2 kDa
Human ROR2, His Tag (RO2-H52E5) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # NP_004551.1).
-20°C
ROR2
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens