Learn More
ACROBiosystems Human S100A6 Protein, Tag Free
Marke: ACROBiosystems S16-H5111-100UG
Weitere Details : Poids : 0.00001kg
Beschreibung
S100 calcium binding protein A6 (S100-A6) is also kown as 2A9, 5B10, CABP, CACY and PRA, is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. Most S100 proteins are disulfide-linked homodimer, and is normally present in cells derived from the neural crest, chondrocytes, macrophages, dendritic cells, etc. S100A6 may function in stimulation of Ca2+ -dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma. S100A6 has been shown to interact with S100B and SUGT1.
This protein carries no "e;tag"e;. The protein has a calculated MW of 10.2 kDa. The protein migrates as 10.2 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH01431 | |
100 μg | |
Met 1 - Gly 90 | |
S100A6,2A9,5B10,CABP,CACY,PRA | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
10.2 kDa | |
Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession # AAH01431). | |
-20°C | |
S100A6 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |