missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human SCARB2 / SR-B2 / LIMPII Protein, Fc Tag

Marque:  ACROBiosystems SC2-H5250-100UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877647

  • 252.20€ / 100µg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Scavenger receptor class B member 2 (SCARB2) is also known as Lysosome membrane protein 2 (LIMP2), 85 kDa lysosomal membrane sialoglycoprotein (LGP85), CD36 antigen-like 2 (CD36L2, LIMP2), which belongs to the CD36 family. SCARB2 acts as a lysosomal receptor for glucosylceramidase (GBA) targeting.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.6 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH21892
HEK293
Human
72.6 kDa
100 µg
Arg 27 - Thr 432
SCARB2,CD36L2,LIMP2,LIMPII
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (HEK293). It contains AA Arg 27 - Thr 432 (Accession # AAH21892).
-20°C
SCARB2
Fc Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions