missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human SECTM1 Protein, His Tag
302.42€ - 2020.12€
Spécification
Numéro d’adhésion | AAH17716 |
---|---|
Poids moléculaire | 13.5 kDa |
Source | Human SECTM1, His Tag (SE1-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 29 - Gly 145 (Accession # AAH17716). |
Immunogène | Gln 29 - Gly 145 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867667
|
ACROBiosystems
SE1-H5227-100UG |
100 μg |
302.42€
100µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877667
|
ACROBiosystems
SE1-H5227-1MG |
1 mg |
2020.12€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Secreted and transmembrane protein 1 (SECTM1) is also known as Protein K-12, which is belongs to the SECTM family, which is Inducted by IFNG/IFN-gamma (at protein level). SECTM1 is detected at the highest levels in peripheral blood leukocytes and breast cancer cell lines. SECTM1 may be involved in thymocyte signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.5 kDa. The protein migrates as 18-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH17716 | |
Human SECTM1, His Tag (SE1-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 29 - Gly 145 (Accession # AAH17716). | |
-20°C | |
SECTM1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
13.5 kDa | |
Gln 29 - Gly 145 | |
SECTM1,K12 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |