Learn More
ACROBiosystems Human Semaphorin 4D / SEMA4D / CD100 Protein, Fc Tag
314.52€ - 2153.18€
Spécification
Numéro d’adhésion | AAH54500 |
---|---|
Poids moléculaire | 105.4 kDa |
Source | Human SEMA4D, Fc Tag (CD0-H5257) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Arg 734 (Accession # AAH54500). |
Immunogène | Met 22 - Arg 734 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876127
|
ACROBiosystems
CD0-H5257-1MG |
1 mg |
2153.18€
1mg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866127
|
ACROBiosystems
CD0-H5257-100UG |
100 μg |
314.52€
100µg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Cluster of Differentiation 100 (CD100) is also known as Semaphorin-4D (SEMA4D), is a single-pass type I membrane protein which belongs to the semaphorin family, and is a human 150-kDa homodimer expressed at the surface of most hemopoietic cells. Semaphorin genes encode soluble and membrane-bound proteins, most of which have been shown to act as chemorepellents on growth cone guidance. CD100 is discrete, as it is a transmembrane leukocyte surface molecule that can also exist in a soluble form. Semaphorin 4D (Sema 4D) is an axon guidance molecule which is secreted by oligodendrocytes and induces growth cone collapse in the central nervous system. By binding plexin B1 receptor it functions as an R-Ras GTPase-activating protein (GAP) and repels axon growth cones in both the mature central nervous system. In the immune system, CD100 binds CD72 to activate B cells and dendritic cells, though much about this interaction is still under investigation. It is involved in oligodendrogenesis during development and during recovery from ischemic injury.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 105.4 kDa. The protein migrates as 125-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH54500 | |
Human SEMA4D, Fc Tag (CD0-H5257) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Arg 734 (Accession # AAH54500). | |
-20°C | |
Semaphorin 4 D | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
105.4 kDa | |
Met 22 - Arg 734 | |
SEMA4D,C9orf164,CD100,FLJ33485,FLJ34282,FLJ39737,FLJ46484,M-sema-G,MGC169138,MGC169141,SEMAJ,coll-4,Semaphorin-4 D | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |