Learn More
Abnova™ Human SEPT12 Partial ORF (AAH35619, 259 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00124404-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).[supplied by OMIM]
Sequence: NGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEFSpécification
AAH35619 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEF | |
RUO | |
SEPT12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
124404 | |
SEPT12 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25410 | |
SEPT12 | |
Recombinant | |
wheat germ expression system |