Learn More
ACROBiosystems Human Siglec-5 / CD170 Protein, Fc Tag
Marque: ACROBiosystems SI5-H5250-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Sialic acid-binding Ig-like lectin 5 (SIGLEC5) is also known as CD33 antigen-like 2 (CD33L2), Obesity-binding protein 2 (OB-BP2), CD antigen CD170, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. SIGLEC5 / CD170 is expressed by monocytic / myeloid lineage cells. SIGLEC-5 / CD33-L2 binds equally to alpha-2,3-linked and alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH29896 | |
73.0 kDa | |
100 µg | |
Glu 17 - Thr 434 | |
SIGLEC5,OB-BP2,CD170,CD33L2 | |
Unconjugated | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human Siglec-5, Fc Tag (SI5-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 17 - Thr 434 (Accession # AAH29896). | |
-20°C | |
Siglec-5 | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |