missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A40 Full-length ORF (NP_061331.2, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00055972-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16152003

  • 335.00€ / 10µg
Expédition estimée: 31-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

SLC25A40 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]

Sequence: MDPETRGQEIIKVTPLQQMLASCTGAILTSVIVTPLDVVKIRLQAQNNPLPKGKCFVYSNGLMDHLCVCEEGGNKLWYKKPGNFQGTLDAFFKIIRNEGIKSLWSGLPPTLVMAVPATVIYFTCYDQLSALLRSKLGENETCIPIVAGIVARFGAVTVISPLELIRTKMQSKKFSYVELHRFVSKKVSEDGWISLWRGWAPTVLRDVPFSAMYWYNYEILKKWLCEKSGLYEPTFMINFTSGALSGSFAAVATLPFDVVKTQKQTQLWTYESHKISMPLHMSTWIIMKNIVAKNGFSGLFSGLIPRLIKIAPACAIMISTYEFGKAFFQKQNVRRQQY
Spécification

Spécification

NP_061331.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MCFP
SLC25A40
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55972
SLC25A40 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDPETRGQEIIKVTPLQQMLASCTGAILTSVIVTPLDVVKIRLQAQNNPLPKGKCFVYSNGLMDHLCVCEEGGNKLWYKKPGNFQGTLDAFFKIIRNEGIKSLWSGLPPTLVMAVPATVIYFTCYDQLSALLRSKLGENETCIPIVAGIVARFGAVTVISPLELIRTKMQSKKFSYVELHRFVSKKVSEDGWISLWRGWAPTVLRDVPFSAMYWYNYEILKKWLCEKSGLYEPTFMINFTSGALSGSFAAVATLPFDVVKTQKQTQLWTYESHKISMPLHMSTWIIMKNIVAKNGFSGLFSGLIPRLIKIAPACAIMISTYEFGKAFFQKQNVRRQQY
RUO
SLC25A40
Wheat Germ (in vitro)
GST
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions