missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC35A3 Partial ORF (NP_036375, 61 a.a. - 113 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00023443-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: DSKCSLRALNRVLHDEILNKPMETLKLAIPSGIYTLQNNLLYVALSNLDAATYSpécification
NP_036375 | |
Liquid | |
23443 | |
SLC35A3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781P1297 | |
SLC35A3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.57kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DSKCSLRALNRVLHDEILNKPMETLKLAIPSGIYTLQNNLLYVALSNLDAATY | |
RUO | |
SLC35A3 | |
Wheat Germ (in vitro) | |
GST |