missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human SLITRK6 Protein, His Tag

Marque:  ACROBiosystems SL6-H5226-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15867707

  • 1815.83€ / 1mg
Date d’expédition estimée: 10-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

SLIT and NTRK-like protein 6 (SLITRK6) belongs to the SLITRK family. SLITRK6 contains 11 LRR (leucine-rich) repeats, two LRRCT domains and two LRRNT domains. In adult brain, SLITRK6 is highly expressed in putamen and also is expressed in adult, fetal lung and fetal liver. SLITRK6 is also expressed at high levels in some brain tumors. Overexpression of SLITRK6 inhibites neurite outgrowth in cultured neurons.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 65.6 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE).
Spécification

Spécification

AAI01071
HEK293
Human
65.6 kDa
1 mg
Ser 27 - Ser 608
SLITRK6
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>90% as determined by SDS-PAGE
Human SLITRK6, His Tag (SL6-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Ser 608 (Accession # AAI01071).
-20°C
SLITRK6
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions