Learn More
ACROBiosystems Human SOD1 / Cu-Zn SOD Protein, His Tag
Marque: ACROBiosystems SO1-H5148-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Superoxide dismutase [Cu-Zn] (SOD1) is also known as superoxide dismutase 1 (hSod1), an enzyme that in humans is encoded by the SOD1 gene, located on chromosome 21. SOD1 can bind copper and zinc ions and is one of three superoxide dismutases responsible for destroying free superoxide radicals in the body. The encoded isozyme (SOD1) is a soluble cytoplasmic and mitochondrial intermembrane space protein, acting as a homodimer to convert naturally occurring, but harmful, superoxide radicals to molecular oxygen and hydrogen peroxide. Furthermore, the mutations of SOD1 gene can result in a neurodegenerative disorder affecting upper motor neurons in the brain and lower motor neurons in the brain stem and spinal cord.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).Spécification
AAH01034 | |
1 mg | |
Ala 2 - Gln 154 | |
SOD1,Superoxide dismutase 1,hSod1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>97% as determined by SDS-PAGE |
16.8 kDa | |
Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession # P60052-1). | |
-20°C | |
SOD1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
E.coli | |
Other Recombinant Proteins |