missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human SOD2 / Mn-SOD Protein, His Tag

sur devis

Spécification

Numéro d’adhésion AAH12423
Poids moléculaire 23.2 kDa
Source Human SOD2, His Tag (SO2-H5126) is expressed from E.coli cells. It contains AA Lys 25 - Lys 222 (Accession # AAH12423).
Immunogène Lys 25 - Lys 222
Conditions de stockage -20°C
Consulter d'autres spécifications

Description

Description

Superoxide dismutase [Mn], mitochondrial (SOD2),a member of the iron/manganese superoxide dismutase family, can form a homotetramer and bind one manganese ion per subunit. The expression of SOD2 is regulated by KRIT1. Mutations in SOD2 gene have been associated with idiopathic cardiomyopathy (IDC), sporadic motor neuron disease, and cancer. Furthermore, Mn-SOD (SOD2) activity is essential to achieve optimal training-induced protection against both ischemia/reperfusion(IR)-induced cardiac arrhythmias and infarction.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE).
Spécification

Spécification

AAH12423
Human SOD2, His Tag (SO2-H5126) is expressed from E.coli cells. It contains AA Lys 25 - Lys 222 (Accession # AAH12423).
-20°C
SOD2
Human
PBS, pH7.4
E.coli
Other Recombinant Proteins
23.2 kDa
Lys 25 - Lys 222
SOD2,IPOB,MNSOD
Unconjugated
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications