missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human SOD2 / Mn-SOD Protein, His Tag

Marque:  ACROBiosystems SO2-H5126-100UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877717

  • 199.74€ / 100µg
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Superoxide dismutase [Mn], mitochondrial (SOD2),a member of the iron/manganese superoxide dismutase family, can form a homotetramer and bind one manganese ion per subunit. The expression of SOD2 is regulated by KRIT1. Mutations in SOD2 gene have been associated with idiopathic cardiomyopathy (IDC), sporadic motor neuron disease, and cancer. Furthermore, Mn-SOD (SOD2) activity is essential to achieve optimal training-induced protection against both ischemia/reperfusion(IR)-induced cardiac arrhythmias and infarction.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE).
Spécification

Spécification

AAH12423
E.coli
Human
23.2 kDa
100 µg
Lys 25 - Lys 222
SOD2,IPOB,MNSOD
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human SOD2, His Tag (SO2-H5126) is expressed from E.coli cells. It contains AA Lys 25 - Lys 222 (Accession # AAH12423).
-20°C
SOD2
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions