missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SUMF1 Partial ORF (NP_877437.2, 89 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00285362-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122757

  • 508.00€ / 25µg
Expédition estimée: 07-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes an enzyme that catalyzes the hydrolysis of sulfate esters by oxidizing a cysteine residue in the substrate sulfatase to an active site 3-oxoalanine residue, which is also known as C-alpha-formylglycine. Mutations in this gene cause multiple sulfatase deficiency, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: SKMVPIPAGVFTMGTDDPQIKQDGEAPARRVTIDAFYMDAYEVSNTEFEKFVNSTGYLTEAEKFGDSFVFEGMLSEQVKTNIQQAVAAAPWWLPVKGANW
Spécification

Spécification

NP_877437.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SKMVPIPAGVFTMGTDDPQIKQDGEAPARRVTIDAFYMDAYEVSNTEFEKFVNSTGYLTEAEKFGDSFVFEGMLSEQVKTNIQQAVAAAPWWLPVKGANW
RUO
SUMF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
285362
SUMF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AAPA3037/FGE/MGC131853/MGC150436
SUMF1
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions