missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human TACI/TNFRSF13B Protein, Fc Tag

sur devis

Spécification

Poids moléculaire 44.6 kDa
Source Human TACI/TNFRSF13B, Fc Tag (TAI-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1).
Immunogène Ser 2 - Thr 166
Conditions de stockage -20°C
Alias de gène CD267,TACI,TNFRSF13B
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15867737
Afficher les documents Détails de la promotion
ACROBiosystems
TAI-H5256-1MG
1 mg Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
15857737
Afficher les documents Détails de la promotion
ACROBiosystems
TAI-H5256-100UG
100 µg Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
Description

Description

Transmembrane activator and CAML interactor (TACI), also known as tumor necrosis factor receptor superfamily member 13B (TNFRSF13B). It was originally discovered because of its ability to interact with calcium-modulator and cyclophilin ligand (CAML). TACI was later found to play a crucial role in humoral immunity by interacting with two members of the TNF family: BAFF and APRIL. The present study demonstrated that, in NSCLC, a proliferation-inducing ligand (APRIL), B-cell maturation antigen (BCMA)and transmembrane activator and CAML interactor (TACI) proteins are abnormally expressed by immunohistochemistry, reverse transcription-quantitative polymerase chain reaction and western blotting. In addition, the expression of APRIL, BCMA and TACI were observed to be involved in extracellular signal-regulated kinase (ERK)1/2 activation in A549 cells.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.6 kDa. The protein migrates as 33-34 kDa, 35 kDa and 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

44.6 kDa
Ser 2 - Thr 166
CD267,TACI,TNFRSF13B
Unconjugated
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>90% as determined by reduced SDS-PAGE
Human TACI/TNFRSF13B, Fc Tag (TAI-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1).
-20°C
TACI
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens
Vidéos
FDS
Documents

Documents

Product Certifications